4-Fluor-2-iodtoluol

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Fluor-3-iod-4-methylbenzol, 4-Fluor-2-iod-1-methylbenzol

TL00300.1 TL00300.10 TL00300.50
MAYBTL00300.1EA 0 EUR
MAYBTL00300.1 MAYBTL00300.10 MAYBTL00300.50
4-Fluor-2-iodtoluol
4-Fluor-2-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236.03 g/mol
MDL: MFCD00013709
CAS-Nummer: 13194-67-7
EINECS: 236-153-7
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR