α,α,α-Trifluor-2-iodtoluol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-Iodobenzotrifluoride, α,α,α-Trifluoro-o-iodotoluene, 1-Iodo-2-(trifluoromethyl)benzene

001194.25G 001194.100G
FLUO001194.25GEA 47.6 EUR
FLUO001194.25G FLUO001194.100G
α,α,α-Trifluor-2-iodtoluol
α,α,α-Trifluor-2-iodtoluol
Formel: C₇H₄F₃I
Molecular Weight: 272,01 g/mol
Siedepunkt: 197 °C (1013 hPa)
Dichte: 1,896 g/cm³ (26 °C)
MDL: MFCD00001041
CAS-Nummer: 444-29-1
EINECS: 207-147-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR