2-Fluor-5-nitrotoluol

Lieferant: Thermo Scientific

Synonyme: 1-Fluoro-2-methyl-4-nitrobenzene

SB01247.1 SB01247.10 SB01247.50
MAYBSB01247.1EA 0 EUR
MAYBSB01247.1 MAYBSB01247.10 MAYBSB01247.50
2-Fluor-5-nitrotoluol
2-Fluor-5-nitrotoluol
Formel: C₇H₆FNO₂
Molecular Weight: 155,13 g/mol
Siedepunkt: 230 °C (1013 hPa)
Schmelzpunkt: 40 °C
MDL: MFCD00007284
CAS-Nummer: 455-88-9
EINECS: 207-251-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR