2-Fluor-4-nitrotoluol
Lieferant: FLUOROCHEM
001186-25G
001186-100G
001186-500G
FLUO001186-25GEA
34.1
EUR
FLUO001186-25G
FLUO001186-100G
FLUO001186-500G
2-Fluor-4-nitrotoluol
2-Fluor-4-nitrotoluol
Formel:
C₇H₆FNO₂ Molecular Weight: 155.13 g/mol Siedepunkt: 235 Schmelzpunkt: 35…36 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007199 CAS-Nummer: 1427-07-2 EINECS: 215-845-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...