4-Fluor-2-methylphenol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
5-Fluoro-2-hydroxytoluene, 1-Fluoro-4-hydroxy-3-methylbenzene
006283-5G
006283-25G
FLUO006283-5GEA
0
EUR
FLUO006283-5G
FLUO006283-25G
4-Fluor-2-methylphenol
4-Fluor-2-methylphenol
Formel:
C₇H₇FO Molecular Weight: 126,13 g/mol Siedepunkt: 84…85 °C (31 mmHg) Schmelzpunkt: 35 °C |
MDL:
MFCD00075088 CAS-Nummer: 452-72-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...