2-Iod-4-nitrotoluol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Iodo-1-methyl-4-nitrobenzene
B21491.06
B21491.14
B21491.06EA
21.4
EUR
B21491.06
B21491.14
2-Iod-4-nitrotoluol ≥97%
2-Iod-4-nitrotoluol
Purity: 97%
CAS No.: 7745-92-8
Molecular Formula: C7H6INO2
Formula Weight: 263.03
MDL No.: MFCD00024329
Formel:
C₇H₆INO₂ Molecular Weight: 263,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024329 CAS-Nummer: 7745-92-8 EINECS: 231-808-3 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...