Benzylamin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Phenylmethylamin, α-Aminotoluol
A10997.0B
A10997.0I
A10997.30
A10997.36
A10997.0BEA
112
EUR
A10997.0B
A10997.0I
A10997.30
A10997.36
Benzylamin ≥98%
Benzylamin
Purity: 98+%
CAS No.: 100-46-9
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 72°(162°F)
Refractive Index: 1.5435
Melting Point: -43°
Boiling Point: 184-185°
Beilstein Reference: 741984
Merck Reference: 14,1125
UN No.: 2735
MDL No.: MFCD00008106
Formel:
(C₆H₅)CH₂NH₂ Molecular Weight: 107,16 g/mol Siedepunkt: 184…185 °C Schmelzpunkt: –43 °C Dichte: 0,982 Flammpunkt: 72 °C (162 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008106 CAS-Nummer: 100-46-9 EINECS: 202-854-1 UN: 2735 ADR: 8,II Merck Index: 14,1125 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...