2-Isopropylanilin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
A14671.14
A14671.22
A14671.36
A14671.14EA
30.7
EUR
A14671.14
A14671.22
A14671.36
2-Isopropylanilin ≥97%
2-Isopropylanilin
Purity: 97%
CAS No.: 643-28-7
Molecular Formula: C9H13N
Formula Weight: 135.21
Flash Point: 95°(203°F)
Refractive Index: 1.5485
Boiling Point: 214-216°
Beilstein Reference: 636283
MDL No.: MFCD00007720
Formel:
C₉H₁₃N Molecular Weight: 135,21 g/mol Siedepunkt: 214…216 °C Dichte: 0,966 Flammpunkt: 95 °C (203 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007720 CAS-Nummer: 643-28-7 EINECS: 211-397-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...