2-Propylanilin
Lieferant: CHEMPUR
|
Achtung
|
JFD03597.250
JFD03597.1
CHMPJFD03597.250EA
0
EUR
CHMPJFD03597.250
MAYBJFD03597.1
2-Propylanilin
2-Propylanilin
Formel:
C₉H₁₃N Molecular Weight: 135,21 g/mol Siedepunkt: 222…224 °C (1013 hPa) Dichte: 0,96 g/cm³ (20 °C) |
MDL:
MFCD00009803 CAS-Nummer: 1821-39-2 EINECS: 217-346-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...