3-tert-Butylanilin

Lieferant: FLUOROCHEM
Achtung

Synonyme: m-tert-Butylanilin

017189-1G 017189-25G 017189-5G
FLUO017189-1GEA 34.1 EUR
FLUO017189-1G FLUO017189-25G FLUO017189-5G
3-tert-Butylanilin
3-tert-Butylanilin
Formel: C₁₀H₁₅N
Molecular Weight: 149.24 g/mol
MDL: MFCD00125078
CAS-Nummer: 5369-19-7
EINECS: 226-361-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR