4-Isopropylanilin ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cumidin
A14646.14
A14646.22
A14646.14EA
48.8
EUR
A14646.14
A14646.22
4-Isopropylanilin ≥99%
4-Isopropylanilin
Purity: 99%
CAS No.: 99-88-7
Molecular Formula: C9H13N
Formula Weight: 135.21
Flash Point: 92°(197°F)
Refractive Index: 1.5430
Melting Point: -63°
Boiling Point: 226-228°
Beilstein Reference: 1930544
Merck Reference: 14,2620
MDL No.: MFCD00007900
Formel:
C₉H₁₃N Molecular Weight: 135,21 g/mol Siedepunkt: 226…228 °C Schmelzpunkt: –63 °C Dichte: 0,989 Flammpunkt: 92 °C (197 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007900 CAS-Nummer: 99-88-7 EINECS: 202-797-2 UN: 1760 ADR: 8,III Merck Index: 14,2620 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...