2-Methylanisol ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Kresolmethylether, o-Methoxytoluol

A13897.18 A13897.30
A13897.18EA 49.4 EUR
A13897.18 A13897.30
2-Methylanisol ≥99%
2-Methylanisol

Purity: 99% CAS No.: 578-58-5 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 51°(123°F) Refractive Index: 1.5170 Boiling Point: 170-172° Beilstein Reference: 1857415 UN No.: 3271 MDL No.: MFCD00008373

Formel: C₈H₁₀O
Molecular Weight: 122,17 g/mol
Siedepunkt: 170…172 °C
Dichte: 0,985
Flammpunkt: 51 °C (123 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008373
CAS-Nummer: 578-58-5
EINECS: 209-426-3
UN: 3271
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR