4-Methylanisol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
p-Methylanisole
A11572.0F
A11572.AE
A11572.AP
A11572.0FEA
261
EUR
A11572.0F
A11572.AE
A11572.AP
4-Methylanisol ≥99%
4-Methylanisol
Purity: 99%
CAS No.: 104-93-8
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 58°(136°F)
Refractive Index: 1.5110
Odour: Aromatic
Melting Point: -50°
Boiling Point: 174-175°
Beilstein Reference: 1237336
UN No.: 3271
MDL No.: MFCD00008413
Formel:
C₈H₁₀O Molecular Weight: 122.17 g/mol Siedepunkt: 174…175 °C Schmelzpunkt: –50 °C Dichte: 0.968 Flammpunkt: 58 °C (136 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008413 CAS-Nummer: 104-93-8 EINECS: 203-253-7 UN: 3271 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...