3-Methylveratrol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,2-Dimethoxy-3-methylbenzol

A11617.06 A11617.14
A11617.06EA 38.1 EUR
A11617.06 A11617.14
3-Methylveratrol ≥98%
3-Methylveratrol

Purity: 98+% CAS No.: 4463-33-6 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 85°(185°F) Refractive Index: 1.5140 Odour: Aromatic Boiling Point: 202-203° Beilstein Reference: 1940962 MDL No.: MFCD00008379

Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Siedepunkt: 202…203 °C
Dichte: 1,024
Flammpunkt: 85 °C (185 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008379
CAS-Nummer: 4463-33-6
EINECS: 224-726-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR