2-Isopropylphenol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Hydroxycumol

A12027.0B A12027.18 A12027.30
A12027.0BEA 144 EUR
A12027.0B A12027.18 A12027.30
2-Isopropylphenol ≥98%
2-Isopropylphenol

Purity: 98% CAS No.: 88-69-7 Molecular Formula: C9H12O Formula Weight: 136.20 Flash Point: 88°(190°F) Physical Form: Clear liquid Density: 1.010 Refractive Index: 1.5270 Appearance: Colourless to pale yellow Melting Point: 14-16° Boiling Point: 212-214° Beilstein Reference: 1363322 UN No.: 2922 MDL No.: MFCD00002224

Formel: C₉H₁₂O
Molecular Weight: 136.19 g/mol
Siedepunkt: 212…214 °C
Schmelzpunkt: 14…16 °C
Dichte: 1.010
Flammpunkt: 88 °C (190 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002224
CAS-Nummer: 88-69-7
EINECS: 201-852-8
UN: 2922
ADR: 8 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR