3-Propylphenol ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-N-Propylphenol

B21266.06 B21266.14
B21266.06EA 121 EUR
B21266.06 B21266.14
3-Propylphenol ≥98%
3-Propylphenol

Purity: 98% CAS No.: 621-27-2 Molecular Formula: C9H12O Formula Weight: 136.19 Refractive Index: 1.5250 Boiling Point: 229-231° UN No.: 3145 MDL No.: MFCD01632130

Formel: C₉H₁₂O
Molecular Weight: 136,19 g/mol
Siedepunkt: 229…231 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632130
CAS-Nummer: 621-27-2
EINECS: 210-675-5
UN: 3145
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR