4-Propylphenol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
p-Propylphenol, 4-n-Propylphenol
A15342.06
A15342.14
A15342.22
A15342.06EA
49
EUR
A15342.06
A15342.14
A15342.22
4-Propylphenol ≥98%
4-Propylphenol
Purity: 98%
CAS No.: 645-56-7
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 106°(222°F)
Refractive Index: 1.5230
Melting Point: 20-22°
Boiling Point: 232°
Beilstein Reference: 1363429
UN No.: 3145
MDL No.: MFCD00002395
Formel:
C₉H₁₂O Molecular Weight: 136,19 g/mol Siedepunkt: 232 °C Schmelzpunkt: 20…22 °C Dichte: 0,983 Flammpunkt: 106 °C (222 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002395 CAS-Nummer: 645-56-7 EINECS: 211-446-2 UN: 3145 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...