tert-Pentylbenzol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Methyl-2-phenylbutane, tert-Amylbenzene

L03731.09 L03731.18
L03731.09EA 48.8 EUR
L03731.09 L03731.18
tert-Pentylbenzol ≥97%
tert-Pentylbenzol

Purity: 97% CAS No.: 2049-95-8 Molecular Formula: C11H16 Formula Weight: 148.25 Flash Point: 65°(149°F) Refractive Index: 1.4950 Boiling Point: 188-191° Beilstein Reference: 2039053 MDL No.: MFCD00015187

Formel: C₆H₅C(CH₃)₂CH₂CH₃
Molecular Weight: 148,25 g/mol
Siedepunkt: 188…191 °C
Dichte: 0,870
Flammpunkt: 65 °C (149 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00015187
CAS-Nummer: 2049-95-8
EINECS: 218-076-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7d664f724255033067c5eed3fe116ec92624378&vid=1e8a332b1dfa9928ae6c1555b83331953d191e08&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR