Cumol ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Phenylpropan, Isopropylbenzol

A11864.0F A11864.AK A11864.AP
A11864.0FEA 107 EUR
A11864.0F A11864.AK A11864.AP
Cumol ≥99%
Cumol

Purity: 99% CAS No.: 98-82-8 Molecular Formula: C9H12 Formula Weight: 120.20 Flash Point: 46°(114°F) Physical Form: Liquid Density: 0.864 Refractive Index: 1.4910 Melting Point: -96° Boiling Point: 152-154° Beilstein Reference: 1236613 Merck Reference: 14,2617 UN No.: 1918 MDL No.: MFCD00008881

Formel: C₉H₁₂
Molecular Weight: 120,19 g/mol
Siedepunkt: 152…154 °C
Schmelzpunkt: –96 °C
Dichte: 0,864
Flammpunkt: 46 °C (114 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008881
CAS-Nummer: 98-82-8
EINECS: 202-704-5
UN: 1918
ADR: 3,III
Merck Index: 14,2617

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR