Toluol 99,7 GC, corrected for water content, BAKER ANALYZED® HPLC HPLC/UHPLC, Spectrophotometry, J.T.Baker®
Lieferant: Avantor
Gefahr
Synonyme: Anisen, Methylbenzol, Methylbenzen, Toluen, Retinnaphtha, Phenylmethan
Nitrogen_filled
9351.1000EA 120.95 EUR
9351.1000 9351.5000
Toluol 99,7 GC, corrected for water content, BAKER ANALYZED® HPLC HPLC/UHPLC, Spectrophotometry, J.T.Baker®
Toluol
Formel: C₆H₅CH₃
Molecular Weight: 92.14 g/mol
Siedepunkt: 110.6
Schmelzpunkt: –95 °C
Dichte: 0.867 g/cm³ (20 °C)
Flammpunkt: 4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008512
CAS-Nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM
Bestellen
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use ≥ 99.7 %
Assay (by GC, corrected for water) ≥ 99.7 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤ 0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm ≤ 0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 288 nm ≤ 0.40
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤ 285
Residue after Evaporation ≤ 3.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤ 0.02 %