Octylbenzol ≥99%

Lieferant: Thermo Scientific

Synonyme: 1-Phenyloctane

A12138.14 A12138.22 A12138.36
A12138.14EA 76.2 EUR
A12138.14 A12138.22 A12138.36
Octylbenzol ≥99%
Octylbenzol

Purity: 99% CAS No.: 2189-60-8 Molecular Formula: C14H22 Formula Weight: 190.33 Flash Point: 107°(224°F) Refractive Index: 1.4840 Odour: Mild Melting Point: -36° Boiling Point: 264-265° Beilstein Reference: 1906253 MDL No.: MFCD00009564

Formel: C₁₄H₂₂
Molecular Weight: 190,33 g/mol
Siedepunkt: 264…265 °C
Schmelzpunkt: –36 °C
Dichte: 0,854
Flammpunkt: 107 °C (224 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009564
CAS-Nummer: 2189-60-8
EINECS: 218-582-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR