Pentylbenzol 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Phenylpentane, Amylbenzene
A15734.06
A15734.14
A15734.22
A15734.06EA
28.5
EUR
A15734.06
A15734.14
A15734.22
Pentylbenzol 96%
Pentylbenzol
Purity: 96%
CAS No.: 538-68-1
Molecular Formula: C11H16
Formula Weight: 148.25
Flash Point: 65°(149°F)
Refractive Index: 1.4880
Melting Point: -75°
Boiling Point: 204-205°
Beilstein Reference: 1904359
Merck Reference: 14,600
UN No.: 3082
MDL No.: MFCD00009502
Formel:
C₁₁H₁₆ Molecular Weight: 148.25 g/mol Siedepunkt: 204…205 °C Schmelzpunkt: –75 °C Dichte: 0.863 Flammpunkt: 65 °C (149 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009502 CAS-Nummer: 538-68-1 EINECS: 208-701-5 UN: 3082 ADR: 9 III Merck Index: 13,00604 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...