Tribenzylamin 99+%
Lieferant: Thermo Scientific
|
Achtung
|
156471000
156472500
156470010
ACRO156471000EA
32.8
EUR
ACRO156471000
ACRO156472500
ACRO156470010
Tribenzylamin 99+%
Tribenzylamin
Formel:
N(CH₂C₆H₅)₃ Molecular Weight: 287,4 g/mol Siedepunkt: 380…390 °C (760 mmHg) Schmelzpunkt: 90…94 °C Dichte: 0,9912 g/cm³ (95 °C) Flammpunkt: 65 °C (closed cup) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004773 CAS-Nummer: 620-40-6 EINECS: 210-638-3 ADR: 4.1,III |
Spezifikation Testergebnisse
Appearance | White to cream Powder |
Infrared spectrum | Conforms |
Melting point | 90 to 94 °C |
Water | ≤0.5 % (K.F.) |
GC | ≥99.0 % |
Titration with HClO4 | ≥99.0 % (On anhydrous substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...