2-Methylbiphenyl ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Phenyltoluol
L01312.06
L01312.03
L01312.06EA
196
EUR
L01312.06
L01312.03
2-Methylbiphenyl ≥98%
2-Methylbiphenyl
Purity: 98%
CAS No.: 643-58-3
Molecular Formula: C13H12
Formula Weight: 168.24
Flash Point: 107°(224°F)
Refractive Index: 1.5910
Melting Point: 0°
Boiling Point: 256°
Beilstein Reference: 2039655
MDL No.: MFCD00008517
Formel:
C₁₃H₁₂ Molecular Weight: 168,24 g/mol Siedepunkt: 256 °C Schmelzpunkt: 0 °C Dichte: 1,010 Flammpunkt: 107 °C (224 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008517 CAS-Nummer: 643-58-3 EINECS: 211-400-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...