4-Methylbiphenyl ≥98%
Lieferant: Thermo Scientific
Synonyme:
4-Phenyltoluol
A14702.03
A14702.06
A14702.14
A14702.03EA
26.1
EUR
A14702.03
A14702.06
A14702.14
4-Methylbiphenyl ≥98%
4-Methylbiphenyl
Purity: 98%
CAS No.: 644-08-6
Molecular Formula: C13H12
Formula Weight: 168.24
Flash Point:
Formel:
C₁₃H₁₂ Molecular Weight: 168,24 g/mol Siedepunkt: 267…268 °C Schmelzpunkt: 44…48 °C Dichte: 1,015 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008544 CAS-Nummer: 644-08-6 EINECS: 211-409-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...