α,α'-Dibrom-m-xylol
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
m-Xyloldibromid, 1,3-Bis(brommethyl)benzol, m-Xylylendibromid
NRB01431.250
NRB01431.1
CHMPNRB01431.250EA
0
EUR
CHMPNRB01431.250
MAYBNRB01431.1
α,α'-Dibrom-m-xylol
α,α'-Dibrom-m-xylol
Formel:
C₈H₈Br₂ Molecular Weight: 263,96 g/mol Siedepunkt: 135…140 °C (20 mmHg) Schmelzpunkt: 76…78 °C Dichte: 1,959 g/cm³ (0 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000178 CAS-Nummer: 626-15-3 EINECS: 210-931-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...