α,α,α',α'-Tetrabrom-p-xylol

Lieferant: Thermo Scientific

Synonyme: alpha,alpha,alpha',alpha'-Tetrabromo-p-xylene, p-Xylylene Tetrabromide, 1,4-Bis(dibromomethyl)benzene

BTB13396.1 BTB13396.10
MAYBBTB13396.1EA 0 EUR
MAYBBTB13396.1 MAYBBTB13396.10
α,α,α',α'-Tetrabrom-p-xylol
α,α,α',α'-Tetrabrom-p-xylol
Formel: C₈H₆Br₄
Molecular Weight: 421,75 g/mol
MDL: MFCD00017857
CAS-Nummer: 1592-31-0
EINECS: 216-473-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR