2,5-Dibrom-p-xylol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,5-Dibrom-1,4-dimethylbenzol, 1,4-Dibrom-2,5-dimethylbenzol

L01776.14 L01776.22
L01776.14EA 45.7 EUR
L01776.14 L01776.22
2,5-Dibrom-p-xylol ≥98%
2,5-Dibrom-p-xylol

Purity: 98+% CAS No.: 1074-24-4 Molecular Formula: C8H8Br2 Formula Weight: 263.97 Odour: Odourless Melting Point: 72-74° Boiling Point: 261° Beilstein Reference: 2080433 MDL No.: MFCD00000091

Formel: C₈H₈Br₂
Molecular Weight: 263,96 g/mol
Siedepunkt: 261 °C
Schmelzpunkt: 72…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000091
CAS-Nummer: 1074-24-4
EINECS: 214-038-2
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR