α-Brom-m-xylol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Methylbenzyl bromide
B24731.06
B24731.14
B24731.06EA
32.1
EUR
B24731.06
B24731.14
α-Brom-m-xylol ≥97%
α-Brom-m-xylol
Purity: 97%
CAS No.: 620-13-3
Molecular Formula: C8H9Br
Formula Weight: 185.07
Storage Temperature: 2°C to 8°C
Flash Point: 82°(179°F)
Refractive Index: 1.5690
Boiling Point: 184-185°/340mm
Beilstein Reference: 2076425
Merck Reference: 14,10090
UN No.: 1701
MDL No.: MFCD00000177
Formel:
C₈H₉Br Molecular Weight: 185,06 g/mol Siedepunkt: 184…185 °C (340 mmHg) Schmelzpunkt: 18 °C Dichte: 1,370 Flammpunkt: 82 °C (179 °F) |
MDL:
MFCD00000177 CAS-Nummer: 620-13-3 EINECS: 210-625-2 UN: 1701 ADR: 6.1,II Merck Index: 14,10090 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...