4-Hydroxy-3,5-dimethylbenzaldehyd
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,5-Dimethyl-4-hydroxybenzaldehyde
BTB03250.1
BTB03250.10
BTB03250.50
MAYBBTB03250.1EA
0
EUR
MAYBBTB03250.1
MAYBBTB03250.10
MAYBBTB03250.50
4-Hydroxy-3,5-dimethylbenzaldehyd
4-Hydroxy-3,5-dimethylbenzaldehyd
Formel:
C₉H₁₀O₂ Molecular Weight: 150,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006946 CAS-Nummer: 2233-18-3 EINECS: 218-774-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...