1-Phenyl-2-harnstoff 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Phenylurea, 1-Phenylurea, N-Phenylurea

131020050 131021000 131022500
ACRO131020050EA 23.8 EUR
ACRO131020050 ACRO131021000 ACRO131022500
1-Phenyl-2-harnstoff 97%
1-Phenyl-2-harnstoff
Formel: C₇H₈N₂O
Schmelzpunkt: 143…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007944
CAS-Nummer: 64-10-8
EINECS: 200-576-5

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 143 to 149 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR