Benzo[b]thiophen 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Thianaphthene, 1-Benzothiophene
138620050
138620250
138621000
ACRO138620050EA
32.8
EUR
ACRO138620050
ACRO138620250
ACRO138621000
Benzo[b]thiophen 97%
Benzo[b]thiophen
Formel:
C₈H₆S Molecular Weight: 134,2 g/mol Siedepunkt: 221…222 °C (760 mmHg) Schmelzpunkt: 28…32 °C Dichte: 1,14 g/cm³ Flammpunkt: 110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005864 CAS-Nummer: 95-15-8 EINECS: 202-395-7 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | White to yellow to pink to orange Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 28 to 33 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...