2-Hydroxyiminopropiophenon 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
2-Hydroxyiminopropiophenone, 2-Isonitrosopropiophenone, 1-Phenyl-1,2-propanedione-2-oxime, 1-Phenyl-1,2-propanedione 2-Oxime
205020250
205020100
205020250.EA
58.8
EUR
205020250.
205020100.
2-Hydroxyiminopropiophenon 99%
2-Hydroxyiminopropiophenon
Formel:
C₉H₉NO₂ Schmelzpunkt: 111…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002115 CAS-Nummer: 119-51-7 EINECS: 204-329-2 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystals |
Infrared spectrum | Conforms |
Melting point | 111 to 115 °C |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...