Salicylaldoxim 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Hydroxybenzaldehyde oxime
207400250
207401000
207405000
207400250.EA
45
EUR
207400250.
207401000.
207405000.
Salicylaldoxim 98%
Salicylaldoxim
Formel:
C₇H₇NO₂ Molecular Weight: 137,14 g/mol Schmelzpunkt: 56…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002120 CAS-Nummer: 94-67-7 EINECS: 202-353-8 |
Spezifikation Testergebnisse
Appearance | White to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 54 to 59 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...