2-Methylphenethylamin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-(o-Tolyl)ethylamine
351100010
351100050
ACRO351100010EA
37.9
EUR
ACRO351100010
ACRO351100050
2-Methylphenethylamin 97%
2-Methylphenethylamin
Formel:
C₉H₁₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310827 CAS-Nummer: 55755-16-3 UN: 2735 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5340 to 1.5360 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...