4-Methyl-2-thiophenboronsäure 96
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4-Methylthiophen-2-yl)boronic acid, 4-Methylthiophene-2-boronic acid
AC35960.250
AC35960.1
MAYBAC35960.250EA
0
EUR
MAYBAC35960.250
MAYBAC35960.1
4-Methyl-2-thiophenboronsäure 96
4-Methyl-2-thiopheneboronic acid
Formel:
C₅H₇BO₂S Molecular Weight: 141.99 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01319040 CAS-Nummer: 162607-15-0 |
Spezifikation Testergebnisse
Appearance | White to light beige Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...