Zinkchlorid 1.0 M in Diethylether, AcroSeal®
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Chlorzink, Zinkdichlorid, Zink(II)chlorid
370051000 370058000
370051000.EA 84.9 EUR
370051000. 370058000.
Zinkchlorid 1.0 M in Diethylether, AcroSeal®
Zinkchlorid
Formel: ZnCl₂
Molecular Weight: 136.3 g/mol
Siedepunkt: 732
Schmelzpunkt: 283
Flammpunkt: –21 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00011295
CAS-Nummer: 7646-85-7
EINECS: 231-592-0
UN: 2924
ADR: 3 I
Merck Index: 13,10185
Bestellen
Spezifikation Testergebnisse
Appearance Colorless to light yellow to light blue-grey Solution (May form two phases which can recombine on cooling. May develop some turbidity or precipitate)
Titration Argentometric 0.9 to 1.1 M
Water ≤0.2 % (Coulometric)