Galenit natürlich vorkommendes Mineral ∼ 1-2 in
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
42546.KF
42546.KJ
42546.KFEA
31.3
EUR
42546.KF
42546.KJ
Galenit natürlich vorkommendes Mineral ∼ 1-2 in
Galenit
Chief ore of lead
- Sp. Gr. 7.4-7.6
Soluble in strong HNO3, in excess of hot HCl
Formel:
̍PbS Molecular Weight: 239,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016280 CAS-Nummer: 1314-87-0 EINECS: 215-246-6 UN: 3077 ADR: 9,III Merck Index: 12,05445 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...