N-Phenyl-p-toluidin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Methyldiphenylamine
44560.09
44560.18
44560.09EA
58.2
EUR
44560.09
44560.18
N-Phenyl-p-toluidin ≥98%
N-Phenyl-p-toluidin
Purity: 98%
CAS No.: 620-84-8
Molecular Formula: C13H13N
Formula Weight: 183.25
Flash Point: 155°(311°F)
Physical Form: Crystalline
Melting Point: 85-86°
Boiling Point: 334°
MDL No.: MFCD00092921
Formel:
C₁₃H₁₃N Molecular Weight: 183,25 g/mol Siedepunkt: 334 °C Schmelzpunkt: 85…86 °C Flammpunkt: 155 °C (311 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092921 CAS-Nummer: 620-84-8 EINECS: 210-655-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...