Salicylaldoxim ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Hydroxybenzaldehyde oxime
A11080.14
A11080.22
A11080.36
A11080.14EA
36.8
EUR
A11080.14
A11080.22
A11080.36
Salicylaldoxim ≥98%
Salicylaldoxim
Purity: 98%
CAS No.: 94-67-7
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Melting Point: 55-59°
Beilstein Reference: 1859881
Merck Reference: 14,8327
MDL No.: MFCD00002120
Formel:
C₇H₇NO₂ Molecular Weight: 137,14 g/mol Schmelzpunkt: 55…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002120 CAS-Nummer: 94-67-7 EINECS: 202-353-8 Merck Index: 14,8327 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...