4-Brom-2-thiophencarbaldehyd 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde
A13304.06
A13304.22
A13304.14
AB114314.0100
A13304.06EA
27.7
EUR
A13304.06
A13304.22
A13304.14
ABCRAB114314.0100
4-Brom-2-thiophencarbaldehyd 96%
4-Brom-2-thiophencarbaldehyd
Purity: 96%
CAS No.: 18791-75-8
Molecular Formula: C5H3BrOS
Formula Weight: 191.05
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₅H₃BrOS Molecular Weight: 191.05 g/mol Siedepunkt: 114…115 °C (11 mmHg) Schmelzpunkt: 44…48 °C Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00005431 CAS-Nummer: 18791-75-8 EINECS: 242-577-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...