L(+)-Citrullin ≥98%
Lieferant: Thermo Scientific
Synonyme:
H-Cit-OH
A13316.09
A13316.18
A13316.09EA
42.7
EUR
A13316.09
A13316.18
L(+)-Citrullin ≥98%
L(+)-Citrullin
Purity: 98%
CAS No.: 372-75-8
Molecular Formula: C6H13N3O3
Formula Weight: 175.19
Melting Point: 212-214° dec.
Beilstein Reference: 6055157
Merck Reference: 14,2331
Optical Rotation: +22° (c=2 in 1N HCl)
MDL No.: MFCD00064397
Formel:
C₆H₁₃N₃O₃ Molecular Weight: 175,19 g/mol Schmelzpunkt: 212…214 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064397 CAS-Nummer: 372-75-8 EINECS: 206-759-6 Merck Index: 14,2331 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...