Perfluor(methylcyclohexan) ≥94%
Lieferant: Thermo Fisher Scientific
A14233.22
A14233.14
A14233.22EA
179
EUR
A14233.22
A14233.14
Perfluor(methylcyclohexan) ≥94%
Perfluor(methylcyclohexan)
Purity: 94%
CAS No.: 355-02-2
Molecular Formula: C7F14
Formula Weight: 350.06
Refractive Index: 1.3 00
Odour: Odourless
Boiling Point: 75-76°
Beilstein Reference: 1915981
MDL No.: MFCD00001468
Formel:
C₇F₁₄ Siedepunkt: 75…76 °C Schmelzpunkt: –70 °C Dichte: 1,800 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001468 CAS-Nummer: 355-02-2 EINECS: 206-573-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...