Metanilgelb
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
A17527.18 A17527.30 A17527.PG
A17527.18EA 26.1 EUR
A17527.18 A17527.30 A17527.PG
Metanilgelb
Metanilgelb
CAS No.: 587-98-4 Molecular Formula: C18H14N3NaO3S Formula Weight: 375.38 Odour: Odourless pH: ~10 Beilstein Reference: 3831568 Merck Reference: 14,5928 MDL No.: MFCD00007486
Formel: C₁₈H₁₄N₃NaO₃S
Molecular Weight: 375,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007486
CAS-Nummer: 587-98-4
EINECS: 209-608-2
Merck Index: 14,5928