3-Methyl-2-thenoesäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Methylthiophene-2-carboxylic acid
A19194.06
A19194.14
A19194.22
A19194.06EA
39.8
EUR
A19194.06
A19194.14
A19194.22
3-Methyl-2-thenoesäure ≥98%
3-Methyl-2-thenoesäure
Purity: 98%
CAS No.: 23806-24-8
Molecular Formula: C6H6O2S
Formula Weight: 142.18
Melting Point: 144-149°
Beilstein Reference: 116184
Formel:
C₆H₆O₂S Molecular Weight: 142,18 g/mol Schmelzpunkt: 144…149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005438 CAS-Nummer: 23806-24-8 EINECS: 245-894-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...