2-Brom-6-methoxynaphthalin ≥98%
Lieferant: Thermo Scientific
A19450.09
A19450.18
AB125719.0010
A19450.09EA
41
EUR
A19450.09
A19450.18
ABCRAB125719.0010
2-Brom-6-methoxynaphthalin ≥98%
2-Bromo-6-methoxynaphthalene
Purity: 98%
CAS No.: 5111-65-9
Molecular Formula: C11H9BrO
Formula Weight: 237.10
Melting Point: 105-110°
Boiling Point: 160-164°/3mm
MDL No.: MFCD00004062
Formel:
C₁₁H₉BrO Molecular Weight: 237,1 g/mol Siedepunkt: 160…164 °C (3 mmHg) Schmelzpunkt: 105…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004062 CAS-Nummer: 5111-65-9 EINECS: 225-837-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...