Methyl-3-ethoxy-2-thiophencarboxylat ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 3-ethoxythiophene-2-carboxylate

B20860.03 B20860.06
B20860.03EA 59.7 EUR
B20860.03 B20860.06
Methyl-3-ethoxy-2-thiophencarboxylat ≥97%
Methyl-3-ethoxy-2-thiophencarboxylat

Purity: 97% CAS No.: 139926-22-0 Molecular Formula: C8H10O3S Formula Weight: 186.23 Melting Point: 72-74° MDL No.: MFCD00102116

Formel: C₈H₁₀O₃S
Molecular Weight: 186,23 g/mol
Schmelzpunkt: 72…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00102116
CAS-Nummer: 139926-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR