Fmoc-Pro-OH ≥98%

Lieferant: Thermo Scientific

Synonyme: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline

B21081.06 B21081.14
B21081.06EA 47.4 EUR
B21081.06 B21081.14
Fmoc-Pro-OH ≥98%
Fmoc-Pro-OH

Purity: 98% CAS No.: 71989-31-6 Molecular Formula: C20H19NO4 Formula Weight: 337.38 Storage Temperature: 2°C to 8°C Melting Point: 113-117° Beilstein Reference: 3596735 Optical Rotation: -40° (c=1 in ethyl acetate) MDL No.: MFCD00037122

Formel: C₂₀H₁₉NO₄
Molecular Weight: 337,38 g/mol
Schmelzpunkt: 113…117 °C
MDL: MFCD00037122
CAS-Nummer: 71989-31-6
EINECS: 276-259-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR