Desylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Chloro-2-phenylacetophenone

B21778.06 B21778.14 B21778.22
B21778.06EA 35.1 EUR
B21778.06 B21778.14 B21778.22
Desylchlorid ≥98%
Desylchlorid

Purity: 98% CAS No.: 447-31-4 Molecular Formula: C14H11ClO Formula Weight: 230.69 Melting Point: 64-69° Beilstein Reference: 744474 UN No.: 3261 MDL No.: MFCD00000858

Formel: C₁₄H₁₁ClO
Molecular Weight: 230,69 g/mol
Schmelzpunkt: 64…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000858
CAS-Nummer: 447-31-4
EINECS: 207-181-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR