2-Benzo[b]thiophenboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Benzo[b]thiophen-2-ylboronic acid, 2-Benzothienylboronic acid

B22835.03 B22835.06 B22835.14
B22835.03EA 34.1 EUR
B22835.03 B22835.06 B22835.14
2-Benzo[b]thiophenboronsäure ≥98%
2-Benzo[b]thiophenboronsäure

Purity: 98% CAS No.: 98437-23-1 Molecular Formula: C8H7BO2S Formula Weight: 178.02 Melting Point: ca 255° Beilstein Reference: 1637924

Formel: C₈H₇BO₂S
Molecular Weight: 178.02 g/mol
Schmelzpunkt: ∼ 255 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075674
CAS-Nummer: 98437-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR